Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0080

Sigma-Aldrich

CJB 090 dihydrochloride hydrate

≥98% (HPLC)

Synonym(s):

N-(4-(4-(2,3-dichlorophenyl)piperazin-1-yl)butyl)-4-(pyridin-2-yl)benzamide dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28Cl2N4O · 2HCl · xH2O
CAS Number:
Molecular Weight:
556.35 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥18 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.Cl.Clc1cccc(N2CCN(CCCCNC(=O)c3ccc(cc3)-c4ccccn4)CC2)c1Cl

InChI

1S/C26H28Cl2N4O.2ClH.H2O/c27-22-6-5-8-24(25(22)28)32-18-16-31(17-19-32)15-4-3-14-30-26(33)21-11-9-20(10-12-21)23-7-1-2-13-29-23;;;/h1-2,5-13H,3-4,14-19H2,(H,30,33);2*1H;1H2

InChI key

MEROVXNAYIISBI-UHFFFAOYSA-N

Biochem/physiol Actions

CJB 090 is a Dopamine D3 receptor partial agonist. CJB 090 was found to attenuate the discriminative stimulus (DS) effects of cocaine while showing no cocaine-like effects when tested alone, and has also been shown to reduce methamphetamine self-administration.
Dopamine D3 receptor partial agonist

Other Notes

Dessicate

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service