Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0078

Sigma-Aldrich

1-Naphthohydroxamic Acid

≥98% (HPLC)

Synonym(s):

N-Hydroxynaphthalene-1-carboxamide, NSC 57457, a-Naphthohydroxamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO2
CAS Number:
Molecular Weight:
187.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

ONC(=O)c1cccc2ccccc12

InChI

1S/C11H9NO2/c13-11(12-14)10-7-3-5-8-4-1-2-6-9(8)10/h1-7,14H,(H,12,13)

InChI key

JRZGPWOEHDOVMC-UHFFFAOYSA-N

Biochem/physiol Actions

1-Naphthohydroxamic Acid is a cell-permeable, potent, and selective histone deacetylase 8 (HDAC8) inhibitor. 1-Naphthohydroxamic acid selectively inhibits HDAC8 (IC50 = 14 μM) over class I HDAC1 and class II HDAC6 (IC50 >100 μM). Treatment of cells with 1-Naphthohydroxamic acid does not increase neither global histone H4 nor tubulin acetylation . Also in contrast to the pan-HDAC inhibitors the compound is not reducing total intracellular HDAC activity.
Cell-permeable, potent, and selective HDAC8 inhibitor

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahesh Chemudupati et al.
Journal of virology, 94(16) (2020-05-29)
Butyrate is an abundant metabolite produced by gut microbiota. While butyrate is a known histone deacetylase inhibitor that activates expression of many genes involved in immune system pathways, its effects on virus infections and on the antiviral type I interferon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service