Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0028

Sigma-Aldrich

FR-171113

≥98% (HPLC)

Synonym(s):

FR-171113, 2-[3-(4-chlorophenyl)-2-[(2,4-dichlorobenzoyl)imino]-4-oxo-5-thiazolidinylidene]-acetic acid, methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H11Cl3N2O4S
CAS Number:
Molecular Weight:
469.73
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥98% (HPLC)

form

powder

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

COC(=O)\C=C1\SC(=N\C(=O)c2ccc(Cl)cc2Cl)\N(C1=O)c3ccc(Cl)cc3

InChI

1S/C19H11Cl3N2O4S/c1-28-16(25)9-15-18(27)24(12-5-2-10(20)3-6-12)19(29-15)23-17(26)13-7-4-11(21)8-14(13)22/h2-9H,1H3/b15-9+,23-19+

InChI key

SDGLYCKTPKZBGI-MSDDKZNZSA-N

Biochem/physiol Actions

FR-171113 is a Thrombin receptor antagonist
FR171113 was the first non-peptide antagonist of the Protease Activated Receptor-1 (PAR1), also termed thrombin receptor. FR171113 inhibits either thrombin or TRAP-6-induced platelet aggregation (IC50 = 290 and 150 nM, respectively). The compound does not affect the protease activity of thrombin or clotting time.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl J May et al.
American journal of physiology. Renal physiology, 317(4), F913-F921 (2019-07-25)
The specific pathogenesis of idiopathic nephrotic syndrome (NS) is poorly understood, and the role of immune mediators remains contentious. However, there is good evidence for the role of a circulating factor, and we recently postulated circulating proteases as candidate factors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service