Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0026

Sigma-Aldrich

4-Phenylbutyryl hydroxamic acid

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥10 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

ONC(=O)CCCc1ccccc1

InChI

1S/C10H13NO2/c12-10(11-13)8-4-7-9-5-2-1-3-6-9/h1-3,5-6,13H,4,7-8H2,(H,11,12)

InChI key

UPHXPXYRKPCXHK-UHFFFAOYSA-N

Biochem/physiol Actions

4-Phenylbutyryl hydroxamic acid is a cell permeable HDAC inhibitor which is about 100 times more potent than parent carboxylic acid among nine HDAC enzymes.
HDAC inhibitor

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service