Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0012

Sigma-Aldrich

SBE 13 hydrochloride

≥98% (HPLC)

Synonym(s):

N-(4-((6-Chloro-3-pyridinyl)methoxy)-3-methoxybenzyl)-N-(2-(3,4-dimethoxyphenyl)ethyl)amine; N-[[4-[(6-Chloro-3-pyridinyl)methoxy]-3-methoxyphenyl]methyl]-3,4-dimethoxy-benzeneethanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27ClN2O4·HCl
CAS Number:
Molecular Weight:
479.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥17 mg/mL

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(CCNCc2ccc(OCc3ccc(Cl)nc3)c(OC)c2)cc1OC

InChI

1S/C24H27ClN2O4.ClH/c1-28-20-7-4-17(12-22(20)29-2)10-11-26-14-18-5-8-21(23(13-18)30-3)31-16-19-6-9-24(25)27-15-19;/h4-9,12-13,15,26H,10-11,14,16H2,1-3H3;1H

InChI key

QBGSVDJLQQXEGG-UHFFFAOYSA-N

application

SBE 13 hydrochloride may be used in cell signaling studies.

Biochem/physiol Actions

SBE 13 is a cell-permeable potent and selective polo-like kinase (Plk1) inhibitor
SBE13 is a cell-permeable highly potent polo-like kinase (Plk1) inhibitor that targets the inactive conformation of the kinase. SBE13 does not effect the activity of Plk2, Plk3, Aurora A kinases.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service