Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00606

Sigma-Aldrich

Dirithromycin

≥96%

Synonym(s):

[9S(R)]-9-Deoxo-11-deoxy-9,11-[imino[2-(2-methoxy)ethylidene]oxy]erthromycin; LY-237216

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H78N2O14
CAS Number:
Molecular Weight:
835.07
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

Quality Level

assay

≥96%

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C4N[C@@H](COCCOC)OC([C@H]4C)[C@]1(C)O

InChI

1S/C42H78N2O14/c1-15-29-42(10,49)37-24(4)32(43-30(56-37)21-52-17-16-50-13)22(2)19-40(8,48)36(58-39-33(45)28(44(11)12)18-23(3)53-39)25(5)34(26(6)38(47)55-29)57-31-20-41(9,51-14)35(46)27(7)54-31/h22-37,39,43,45-46,48-49H,15-21H2,1-14H3/t22-,23-,24+,25+,26-,27+,28+,29-,30-,31+,32+,33-,34+,35+,36-,37-,39+,40-,41+,42-/m1/s1

InChI key

WLOHNSSYAXHWNR-KZYCBHIHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dirithromycin is a macrolide antibiotic. It is a prodrug of erythromycylamine that has outstanding activity against Campylobacter. Dirithromycin is a group 3 agent with respect to its interaction with cytochrome P450 (CYP) isoform 3A4, as it interferes poorly with CYP3A4 in vitro and generally does not alter drug metabolism in vivo.

Packaging

1G

Preparation Note

When dissolving dirithromycin in organic solvents, first purge the solvent with inert gas.

To prepare in aqueous buffer, dirithromycin should first be dissolved in ethanol and then diluted with aqueous buffer. The solution is stable for one day.

Dirithromycin is soluble in ethanol at approximately 20 mg/mL, DMF at approximately 10 mg/mL, and DMSO at approximately 15 mg/mL.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macrolide - induced clinically relevant drug interactions with cytochrome P-450A (CYP) 3A4: an update focused on clarithromycin, azithromycin and dirithromycin.
Westphal JF
British Journal of Clinical Pharmacology, 50(4), 285-295 (2000)
Comparative in vitro activities of new 14-, 15-, and 16-membered macrolides.
Hardy DJ, Hensey DM, Beyer JM, Vojtko C, McDonald EJ, Fernandes PB.
Antimicrobial Agents and Chemotherapy, 32(11), 1710-1719 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service