Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00585

Sigma-Aldrich

1,3-Dipalmitoyl-sn-glycero-2-PE

≥95% (HPLC)

Synonym(s):

1,3-DPPE, 1,3-Dipalmitoyl-glycero-2-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H74NO8P
CAS Number:
Molecular Weight:
691.96
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

assay

≥95% (HPLC)

form

powder

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

O=C(OCC(OP(O)(OCCN)=O)COC(CCCCCCCCCCCCCCC)=O)CCCCCCCCCCCCCCC

InChI

1S/C37H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(46-47(41,42)45-32-31-38)34-44-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34,38H2,1-2H3,(H,41,42)

InChI key

OQEIOUBBMBZINL-UHFFFAOYSA-N

Biochem/physiol Actions

Phospholipids found in biological membranes are known as phosphatidylethanolamines (PEs). PEs are commonly used in the formation of micelles and liposomes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service