Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00514

Sigma-Aldrich

Polystachoside

≥95% (LC/MS-UV)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$318.00

$318.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$318.00

About This Item

Empirical Formula (Hill Notation):
C20H18O11
CAS Number:
Molecular Weight:
434.35
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$318.00


Usually ships in 2 weeks.

Request a Bulk Order

assay

≥95% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]1CO)O)O)Oc2[c](c3c([o]c2c4cc(c(cc4)O)O)cc(cc3O)O)=O

InChI

1S/C20H18O11/c21-6-13-15(26)17(28)20(30-13)31-19-16(27)14-11(25)4-8(22)5-12(14)29-18(19)7-1-2-9(23)10(24)3-7/h1-5,13,15,17,20-26,28H,6H2/t13-,15-,17+,20-/m0/s1

InChI key

BDCDNTVZSILEOY-UXYNSRGZSA-N

General description

Natural product derived from plant source.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service