Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00503

Sigma-Aldrich

3-Propoxypropane-1,2-diol

≥98% (GC)

Synonym(s):

1-O-Propylglycerol, 3-propoxypropylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O3
CAS Number:
Molecular Weight:
134.17
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:

assay

≥98% (GC)

form

liquid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OCC(O)COCCC

InChI

1S/C6H14O3/c1-2-3-9-5-6(8)4-7/h6-8H,2-5H2,1H3

InChI key

ZTKZJXGLCCVMLJ-UHFFFAOYSA-N

General description

3-Propoxypropane-1,2-diol (Glycerol 1-propyl ether, 1-O-Propyl glycerol) is a propyl ether of glycerol.[1] It can be prepared by reacting glycerol with propyl bromide using sodium hydride in dry dimethylformamide.[2]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

242.6 °F

flash_point_c

117 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bidirectional conversion between 3-monochloro-1,2-propanediol and glycidol in course of the procedure of DGF standard methods.
Kaze N, et al.
Journal of the American Oil Chemists' Society, 88(8), 1143-1151 (2011)
Hellwinkel D.
Systematic Nomenclature of Organic Chemistry: A Directory to Comprehension and Application of its Basic Principles, 131-131 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service