Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00471

Sigma-Aldrich

Monotropein

≥98% (HPLC)

Synonym(s):

[1S-(1α, 4aα,7β,7aα)]-1-(β-D-Glucopyranosyloxy)-1,4a,7,7a-tetrahydro-7-hydroxy-7-(hydroxymethyl)cyclopenta[c]pyran-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O11
CAS Number:
Molecular Weight:
390.34
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@@H](O)[C@@](O[C@H]2[C@@]([C@](CO)(O)C=C3)([H])[C@@]3([H])C(C(O)=O)=CO2)([H])O[C@@H]1CO

InChI

1S/C16H22O10/c17-3-7-5-24-14(10-8(7)1-2-16(10,23)6-19)26-15-13(22)12(21)11(20)9(4-18)25-15/h1-3,5,8-15,18-23H,4,6H2/t8-,9-,10-,11-,12+,13-,14+,15+,16+/m1/s1

InChI key

CYYIGLDAAXXQAI-PSWNKMJPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Monotropein is an iridoid glycoside found in the root of Morinda officinalis.

Biochem/physiol Actions

Monotropein has anti-inflammatory effects in vivo due to its ability to inhibit the expression of inflammatory mediators via NF-KB inactivation.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Courtney P Leisner et al.
PloS one, 12(6), e0179417-e0179417 (2017-06-14)
Cultivated blueberry (Vaccinium corymbosum, Vaccinium angustifolium, Vaccinium darrowii, and Vaccinium virgatum) is an economically important fruit crop native to North America and a member of the Ericaceae family. Several species in the Ericaceae family including cranberry, lignonberry, bilberry, and neotropical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service