Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00458

Sigma-Aldrich

Neobavaisoflavone

≥98% (HPLC)

Synonym(s):

7-Hydroxy-3-(4-hydroxy-3-(3-methyl-2-buten-1-yl)phenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O4
CAS Number:
Molecular Weight:
322.35
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC1=CC=C(C(C(C2=CC(CC=C(C)C)=C(O)C=C2)=CO3)=O)C3=C1

InChI

1S/C20H18O4/c1-12(2)3-4-14-9-13(5-8-18(14)22)17-11-24-19-10-15(21)6-7-16(19)20(17)23/h3,5-11,21-22H,4H2,1-2H3

InChI key

OBGPEBYHGIUFBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neobavaisoflavone (NBIF) is an isoflavone found in Psoralea corylifolia.

Biochem/physiol Actions

Neobavaisoflavone has been reported to have anti-inflammatory, anti-cancer, and anti-fungal activities. It also acts as a DNA polymerase and platelet aggregation inhibitor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lishan Zhou et al.
Frontiers in pharmacology, 10, 1589-1589 (2020-03-03)
Psoralea corylifolia L. (PC) is a traditional Chinese herb used to treat yang deficiency of the spleen and kidney in pediatric disease. Our previous studies have found that PC can alleviate the liver oxidative stress of juvenile mice with nonalcoholic
Parham Taslimi
Journal of biochemical and molecular toxicology, 34(11), e22566-e22566 (2020-07-03)
Tyrosinase enzyme is a functional oxidase that is extensively divided in nature. It is the main enzyme in melanin synthesis and is also involved in designating the color of mammalian hair and skin. Additionally, it is accountable for the unfavorable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service