Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00451

Sigma-Aldrich

Jujuboside B

≥98% (HPLC)

Synonym(s):

Jujubopside B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H84O21
CAS Number:
Molecular Weight:
1045.21
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Semen Ziziphus spinosa)

assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@@]12[C@](CC[C@]3(C)[C@]2([H])CC[C@@]4([H])[C@@]3(CO5)C[C@@]65[C@]4([H])[C@@](C)(O)C[C@H](C=C(C)C)O6)([H])C(C)(C)[C@@H](O[C@]7([H])OC[C@H](O)[C@H](O[C@]8([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O[C@@]9([H])[C@H](O)[C@@H](O)[C@H](O)CO9)[C@H]7O[C@@]%10([H])

InChI

1S/C52H84O21/c1-22(2)15-24-16-50(8,63)42-25-9-10-30-48(6)13-12-31(47(4,5)29(48)11-14-49(30,7)51(25)20-52(42,73-24)66-21-51)69-45-41(72-44-38(62)35(59)32(56)23(3)67-44)39(27(55)19-65-45)70-46-40(36(60)34(58)28(17-53)68-46)71-43-37(61)33(57)26(54)18-64-43/h15,23-46,53-63H,9-14,16-21H2,1-8H3/t23-,24-,25+,26+,27-,28+,29-,30+,31-,32-,33-,34+,35+,36-,37+,38+,39-,40+,41+,42-,43-,44-,45-,46-,48-,49+,50-,51-,52-/m0/s1

InChI key

OAVAUZCEOWCYCC-QEOGCQCLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Jujuboside B (JuB) is a saponin isolated from Semen Ziziphus spinosa (SZS) used in Traditional Chinese Medicine.[1]

Biochem/physiol Actions

Jujuboside B is known for its treatment of insomnia and anxiety[2][3] and has been reported to have activities relevant to the cardiovascular system.[4] Jujuboside B inhibits platelet aggregation and is considered as a preventive drug for platelet-based cardiovascular diseases.[5]
Jujuboside B as food supplement, has neurobeneficial effects[6] and could be a potential drug candidate for treating neurodegenerative diseases.[6]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zizyphus jujuba and its active component jujuboside B inhibit platelet aggregation.
Seo E J, et al.
Phytotherapy Research, 27(6), 829-834 (2013)
Ziziphus spinosa seeds for insomnia: a review of chemistry and psychopharmacology.
Shergis J L, et al.
Phytomedicine, 34, 38-43 (2017)
Herbal medicine for anxiety, depression and insomnia.
Liu L, et al.
Current Neuropharmacology, 13(4), 481-493 (2015)
Jujuboside B reduces vascular tension by increasing Ca2+ influx and activating endothelial nitric oxide synthase.
Zhao Y, et al.
PLoS ONE, 11(2), e0149386-e0149386 (2016)
Hypnotic effect of jujubosides from Semen Ziziphi Spinosae.
Cao J X, et al.
Journal of Ethnopharmacology, 130(1), 163-166 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service