Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00431

Sigma-Aldrich

Mollugin

≥98% (HPLC)

Synonym(s):

6-Hydroxy-2,2-dimethyl-2H-naphtho(1,2-b)pyran-5-carboxylic acid methyl ester, Rubimaillin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16O4
CAS Number:
Molecular Weight:
284.31
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC1(C)C=CC(C(C(OC)=O)=C(O)C2=C3C=CC=C2)=C3O1

InChI

1S/C17H16O4/c1-17(2)9-8-12-13(16(19)20-3)14(18)10-6-4-5-7-11(10)15(12)21-17/h4-9,18H,1-3H3

InChI key

VLGATXOTCNBWIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mollugin is a naphthohydroquinone found in the roots of Rubia cordifolia L. that exhibits antimutagenic, antitumor, and antiviral activities.

Biochem/physiol Actions

Mollugin displays inhibitory activity in arachidonic acid- and collagen-induced platelet aggregation. Mollugin has been reported to inhibit cancer cell proliferation, and has other biological activities including anti-inflammatory and apoptotic effects.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service