Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00402

Sigma-Aldrich

β-Glc-PEG3-Azide

≥95%

Synonym(s):

(8-Azido-3,6-dioxaoctyl) β-D-glucopyranoside, 2-{2-[2-azidoethoxy]ethoxy}ethyl β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23N3O8
CAS Number:
Molecular Weight:
337.33
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic (organic)

assay

≥95%

form

liquid

storage temp.

−20°C

SMILES string

OCC1O[C@@H](OCCOCCOCCN=[N+]=[N-])[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H23N3O8/c13-15-14-1-2-20-3-4-21-5-6-22-12-11(19)10(18)9(17)8(7-16)23-12/h8-12,16-19H,1-7H2/t8?,9-,10-,11?,12+/m0/s1

InChI key

ABBZTEUICIJBPU-MUVKAJBYSA-N

General description

β-Glc-PEG3-Azide is a PEGylated glycoside.

Biochem/physiol Actions

β-Glc-PEG3-Azide is a PEGylated glycoside. PEGylated glycosides are used as ligands for Click Chemistry. They may also be important as carbohydrate carriers in glycotherapeutic applications, drug carriers for site-specific delivery, drug enhancers for decreasing toxicity, and solubility and protein enhancers.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service