Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00336

Sigma-Aldrich

Aristolochic acid B

≥97% (HPLC)

Synonym(s):

6-Nitrophenanthro(3,4-d)-1,3-dioxole-5-carboxylic acid, Aristolochic acid II, Noraristolochic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9NO6
CAS Number:
Molecular Weight:
311.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥97% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC(C1=C2C([N+]([O-])=O)=CC3=CC=CC=C3C2=C4OCOC4=C1)=O

InChI

1S/C16H9NO6/c18-16(19)10-6-12-15(23-7-22-12)14-9-4-2-1-3-8(9)5-11(13(10)14)17(20)21/h1-6H,7H2,(H,18,19)

InChI key

MEEXETVZNQYRSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aristolochic acid B or Aristolochic acid II (AA-II) is one of the most abundant secondary metabolites produced in leaves, roots, and stems of Aristolochiaceae family plants. AA-II is structurally similar to Aristolochic acid I (AA-I) with the only difference being the methoxy group at C8.

Application

Aristolochic acid B may have been used to test its fungi-toxic effect on Botrytis cinerea. It may also have been used as a reference standard to study its effects on in vivo metabolism using high-performance liquid chromatography/electrospray ionization mass spectrometry (HPLC/ESI-MS).

Biochem/physiol Actions

Aristolochic acid B inhibits in vitro phospholipid hydrolysis by HSF-PLA2 in a dose-dependent manner.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 Inhalation - Muta. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alena Dedı Ková et al.
Chemical research in toxicology, 33(11), 2804-2818 (2020-09-08)
The plant extract aristolochic acid (AA), containing aristolochic acid I (AAI) and II (AAII) as major components, causes aristolochic acid nephropathy and Balkan endemic nephropathy, unique renal diseases associated with upper urothelial cancer. Differences in the metabolic activation and detoxification
Sivaprasad Attaluri et al.
Nucleic acids research, 38(1), 339-352 (2009-10-27)
Aristolochic acids I and II (AA-I, AA-II) are found in all Aristolochia species. Ingestion of these acids either in the form of herbal remedies or as contaminated wheat flour causes a dose-dependent chronic kidney failure characterized by renal tubulointerstitial fibrosis.
R Melo et al.
Letters in applied microbiology, 68(1), 48-55 (2018-10-17)
The fungitoxic effect of aristolochic acids I and II on mycelial growth and conidial germination of Botrytis cinerea was analysed. Aristolochic acid I had a higher effect on mycelial growth of B. cinerea than aristolochic acid II with IC50 value

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service