Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00318

Sigma-Aldrich

Astragaloside I

≥97% (HPLC)

Synonym(s):

(3β,6α,16β,20R,24S)-3-[(2,3-Di-O-acetyl-β-D-xylopyranosyl)oxy]-20,24-epoxy-16,25-dihydroxy-9,19-cyclolanostan-6-yl β-D-glucopyranoside, Astrasieversianin IV, Cyclosieversioside B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H72O16
CAS Number:
Molecular Weight:
869.04
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

assay

≥97% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)[C@@H](O[C@]2([H])OC[C@@H](O)[C@H](OC(C)=O)[C@H]2OC(C)=O)CC[C@@]3(C4)[C@@]1([H])[C@@H](O[C@]5([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C[C@]6([H])[C@@]34CC[C@]7(C)[C@]([C@]8(C)O[C@H](C(O)(C)C)CC8)([H])[C@@H](O)C[C@@]67C

InChI

1S/C45H72O16/c1-21(47)56-33-24(50)19-55-38(34(33)57-22(2)48)60-28-11-13-45-20-44(45)15-14-41(7)35(43(9)12-10-29(61-43)40(5,6)54)23(49)17-42(41,8)27(44)16-25(36(45)39(28,3)4)58-37-32(53)31(52)30(51)26(18-46)59-37/h23-38,46,49-54H,10-20H2,1-9H3/t23-,24+,25-,26+,27-,28-,29-,30+,31-,32+,33-,34+,35-,36-,37+,38-,41+,42-,43+,44-,45+/m0/s1

InChI key

KXHCYYSIAXMSPA-OOCCOBHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Astragaloside I is a saponin found in the root of Astragalus propinquus, which is known as huáng qí in Traditional Chinese Medicine.

Biochem/physiol Actions

Astragaloside I has been shown to increase gastric pH without changing gastric volume or acid output.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service