Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00309

Sigma-Aldrich

Fmoc-L-Val(i,i-Fru)-OH

≥90% (HPLC)

Synonym(s):

N-α-Fmoc-N-α-(1-deoxy-2,3:4,5-di-O-isopropylidene-β-D-fructopyranos-1-yl)-L-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H39NO9
Molecular Weight:
581.65
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic (organic)

assay

≥90% (HPLC)

form

powder

functional group

Fmoc

storage temp.

−20°C

SMILES string

CC1(C)O[C@@H]2CO[C@]3(CN(C(OCC4C5=C(C6=C4C=CC=C6)C=CC=C5)=O)[C@@H](C(C)C)C(O)=O)C(CC(C)(C)O3)[C@@H]2O1

InChI

1S/C33H41NO8/c1-19(2)27(29(35)36)34(18-33-25(15-31(3,4)42-33)28-26(17-39-33)40-32(5,6)41-28)30(37)38-16-24-22-13-9-7-11-20(22)21-12-8-10-14-23(21)24/h7-14,19,24-28H,15-18H2,1-6H3,(H,35,36)/t25?,26-,27+,28-,33+/m1/s1

InChI key

CASOLNDWJFMLTG-CSDUBWOFSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service