Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SMB00299

Sigma-Aldrich

Fmoc-L-Ser((Ac)3-β-D-GlcNAc)-OH

≥95% (HPLC)

Synonym(s):

Fmoc-L-Ser((Ac)3-β-D-GlcNAc)-OH, N-α-(9-Fluorenylmethyloxycarbonyl)-O-β-(2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl)-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N2O13
CAS Number:
Molecular Weight:
656.63
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

assay

≥95% (HPLC)

form

powder

functional group

Fmoc

storage temp.

−20°C

SMILES string

[R]C[C@H]1O[C@@H](OC[C@@H](NC(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)C(O)=O)[C@H](NC(C)=O)C([R])[C@@H]1[R]

InChI

1S/C32H36N2O13/c1-16(35)33-27-29(46-19(4)38)28(45-18(3)37)26(15-42-17(2)36)47-31(27)43-14-25(30(39)40)34-32(41)44-13-24-22-11-7-5-9-20(22)21-10-6-8-12-23(21)24/h5-12,24-29,31H,13-15H2,1-4H3,(H,33,35)(H,34,41)(H,39,40)/t25-,26+,27+,28+,29+,31+/m0/s1

InChI key

ORICVOOXZDVFIP-ILRBKQRBSA-N

General description

A basic monosaccharide fragment of branched N-linked oligosaccharides found on glycoproteins.

Biochem/physiol Actions

In dynamic protein glycosylation, reversible O-GlcNAc modification of nuclear and cytoplasmic proteins at serine and threonine controls diverse aspects of cell physiology and is conceptually analogous to phosphorylation.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service