Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00284

Sigma-Aldrich

Glucose-UDP-(PEG)6-Fluorescein Conjugate

Synonym(s):

Glucose-UDP-(PEG)6-Fluorescein Conjugate, Glucose-Uridine Diphosphate-(PEG)6-Fluorescein Conjugate, UDP-Glc-(PEG)6-Fluorescein Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

conjugate

fluorescein conjugate

assay

≥95% (HPLC)

form

solid

fluorescence

λex 490 nm; λem 514 nm

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OP(OP([O-])(OC[C@H]2O[C@@H](N(C=C3/C=C/CNC(CCOCCOCCOCCOCCNC(C4=CC=C(C([O-])=O)C(C(C5=C(O6)C=C(O)C=C5)=C7C6=CC(C=C7)=O)=C4)=O)=O)C(NC3=O)=O)[C@H](O)[C@@H]2O)=O)([O-])=O)O[C@H](CO)[C@H]1O.CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC

InChI

1S/C50H60N4O28P2.3C6H15N/c55-24-36-40(59)42(61)44(63)49(80-36)81-84(71,72)82-83(69,70)77-25-37-41(60)43(62)47(79-37)54-23-27(46(65)53-50(54)68)2-1-10-51-38(58)9-12-73-14-16-75-18-19-76-17-15-74-13-11-52-45(64)26-3-6-30(48(66)67)33(20-26)39-31-7-4-28(56)21-34(31)78-35-22-29(57)5-8-32(35)39;3*1-4-7(5-2)6-3/h1-8,20-23,36-37,40-44,47,49,55-56,59-63H,9-19,24-25H2,(H,51,58)(H,52,64)(H,66,67)(H,69,70)(H,71,72)(H,53,65,68);3*4-6H2,1-3H3/b2-1+;;;/t36?,37-,40-,41-,42-,43-,44?,47-,49+;;;/m1.../s1

InChI key

YLXCDZIEXNLUEA-MBSOWBKXSA-N

Biochem/physiol Actions

Glucose-UDP, or UDP-Glucose, is a nucleotide sugar that plays a key role in carbohydrate metabolism. UDP-Glucose is a precursor to glycogen and a key substrate for glycosyltransferases, such as Toxins A and B from Clostridium difficile. Fluorescent analogs of UDP-Glucose can be used for studying glucosyltransferases and clostridial toxins. This fluorescein UDP-Glucose derivative has spectral properties identical to those of FITC conjugates. Compared to SMB00285, this UDP-Glucose derivative has a much longer spacer between the fluorophore and UDP moiety.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service