Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00228

Sigma-Aldrich

Isopongaflavone

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O4
CAS Number:
Molecular Weight:
334.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc2OC(C)(C)C=Cc2c3OC(=CC(=O)c13)c4ccccc4

InChI

1S/C21H18O4/c1-21(2)10-9-14-17(25-21)12-18(23-3)19-15(22)11-16(24-20(14)19)13-7-5-4-6-8-13/h4-12H,1-3H3

InChI key

DPAGRPSAFDXQDN-UHFFFAOYSA-N

General description

Natural product derived from plant source.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service