Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00207

Sigma-Aldrich

Cornuside

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O14
CAS Number:
Molecular Weight:
542.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C24H30O14/c1-3-11-12(4-5-35-21(32)10-6-14(26)17(28)15(27)7-10)13(22(33)34-2)9-36-23(11)38-24-20(31)19(30)18(29)16(8-25)37-24/h3,6-7,9,11-12,16,18-20,23-31H,1,4-5,8H2,2H3/t11-,12+,16+,18+,19-,20+,23+,24-/m0/s1

InChI key

SMTKSCGLXONVGL-ZEIULZLQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Biochem/physiol Actions

Cornuside is a secoiridoid glucoside with anti-inflammatory properties through the downregulation of iNOS and COX-2 via NF-kB inhibition.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Zhao et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 27(11), 845-848 (1992-01-01)
A new bisiridoid glucoside named cornuside was isolated from the fruits of Cornus officinalis Sieb et Zucc. The structure of cornuside has been established by spectroscopic methods (IR, MS, 1HNMR, 13CNMR).
Shun Zong Song et al.
Bioscience, biotechnology, and biochemistry, 75(4), 656-661 (2011-04-23)
This study elucidated the effects of cornuside on carbon tetrachloride (CCl₄)-induced hepatotoxicity. Rats were treated intraperitoneally with 0.5 mL/kg of CCl₄. Sixteen h after CCl₄ treatment, the levels of serum aminotransferases, tumor necrosis factor-α (TNF-α), and lipid peroxidation were significantly
Dae Gill Kang et al.
Biological & pharmaceutical bulletin, 30(9), 1796-1799 (2007-09-11)
Cornuside is a bisiridoid glucoside compound isolated from the fruit of Cornus officinalis SIEB. et ZUCC. The present study was designed to examine the effects of cornuside on expression levels of cytokine-induced proinflammatory and adhesion molecules in the human umbilical
Wang-Lin Jiang et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(4), 266-271 (2010-08-27)
Cornuside is a secoiridoid glucoside isolated from the fruit of Cornus officinalis SIEB. et ZUCC. In this study, we investigated the anti-myocardial ischemia and reperfusion (I/R) injury effects of cornuside in vivo and elucidated the potential mechanism. Rat models of
Dae Gill Kang et al.
Planta medica, 73(14), 1436-1440 (2007-10-11)
Cornuside is a bisiridoid glucoside compound isolated from the fruit of CORNUS OFFICINALIS Sieb. et Zucc. (Cornaceae). In the present study, we investigated the effect of cornuside on vascular tone in rat aortic tissue. Cornuside induced a concentration-dependent relaxation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service