Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00128

Sigma-Aldrich

Dihydroyashabushiketol

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O2
CAS Number:
Molecular Weight:
282.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@@H](CCc1ccccc1)CC(=O)CCc2ccccc2

InChI

1S/C19H22O2/c20-18(13-11-16-7-3-1-4-8-16)15-19(21)14-12-17-9-5-2-6-10-17/h1-10,18,20H,11-15H2/t18-/m0/s1

InChI key

CCNKTMMNRPJQHV-SFHVURJKSA-N

General description

Natural product derived from plant source.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service