Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00122

Sigma-Aldrich

Vincosamide

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N2O8
CAS Number:
Molecular Weight:
498.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2OC=C3[C@@H](C[C@H]4N(CCc5c4[nH]c6ccccc56)C3=O)[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C26H30N2O8/c1-2-12-15-9-18-20-14(13-5-3-4-6-17(13)27-20)7-8-28(18)24(33)16(15)11-34-25(12)36-26-23(32)22(31)21(30)19(10-29)35-26/h2-6,11-12,15,18-19,21-23,25-27,29-32H,1,7-10H2/t12-,15+,18-,19-,21-,22+,23-,25+,26+/m1/s1

InChI key

LBRPLJCNRZUXLS-AZVRXDBZSA-N

General description

Natural product derived from plant source.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samah R Khalil et al.
Gene, 730, 144272-144272 (2019-12-10)
Tilmicosin (Til), an effective macrolide antibiotic, is widely used against respiratory diseases in livestock; however, its treatment is associated with cardiac tissue impairments. In this study, the ethanolic extract of Moringa oleifera (MO) leaves was investigated at two doses (400
Hongqiang Lin et al.
Molecules (Basel, Switzerland), 24(5) (2019-03-15)
With the aim to discuss the similarities and differences of phytochemicals in Moringa oleifera leaves collected from China (CML) and India (IML) in mind, comparative ultra-performance liquid chromatography coupled with quadrupole-time-of-flight mass spectrometry (UPLC-QTOF-MS) analysis was performed in this study.
Carla R F Volobuff et al.
Current pharmaceutical biotechnology, 20(4), 302-308 (2019-02-13)
The genus Psychotria and Palicourea are reported as a source of alkaloids and iridoids, which exhibit biological activities. This study aimed to evaluate antiproliferative and anticholinesterase activities and quantification of the alkaloids of seven species among the genus found in
Zheng Wei Wong et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 94, 1145-1166 (2017-08-23)
Modern medicine has been used to treat myocardial infarction, a subset of cardiovascular diseases, and have been relatively effective but not without adverse effects. Consequently, this issue has stimulated interest in the use of natural products, which may be equally
Samah R Khalil et al.
Ecotoxicology and environmental safety, 200, 110716-110716 (2020-05-26)
This study assessed the potential of Moringa oleifera leaves ethanol extract (MLEE) in attenuating the detrimental effects of cobalt dichloride (CoCl2) on rat liver. Forty rats were assigned to five equal groups: control group, MLEE-treated group, CoCl2-treated group, prophylaxis co-treated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service