Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00034

Sigma-Aldrich

6-Deoxy-8-O-methylrabelomycin

≥95% (LC/MS-ELSD)

Synonym(s):

MM 47755

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16O5
CAS Number:
Molecular Weight:
336.34
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C20H16O5/c1-20(24)8-10-6-7-12-17(15(10)13(21)9-20)19(23)11-4-3-5-14(25-2)16(11)18(12)22/h3-7,24H,8-9H2,1-2H3/t20-/m1/s1

InChI key

XZLGWJORNHETKI-HXUWFJFHSA-N

General description

Natural product derived from fungal source.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakeline Trejos Jiménez et al.
Journal of microbiology (Seoul, Korea), 50(6), 1014-1023 (2013-01-01)
Streptomyces mediolani sp. AC37 was isolated from the root system of higher plant Taxus baccata and produced metabolite identified as (-)-8-O-methyltetrangomycin according to LC/MS/MS analysis. In our screening program for improvements of bioactive secondary metabolites from plant associate streptomycetes, mutation
K Nemoto et al.
The Journal of antibiotics, 41(9), 1253-1259 (1988-09-01)
An assessment of the prophylactic and ameliorative effects of deoxyspergualin (NKT-01), an immunosuppressive agent, was carried out in male MRL/MpJ-lpr/lpr (MRL/1) mice which spontaneously develop lupus-like lesions. When NKT-01 was administered ip daily from the age of either 8 or
MM 47755, a new benz[a]anthracene antibiotic from a streptomycete.
M L Gilpin et al.
The Journal of antibiotics, 42(4), 627-628 (1989-04-01)
Christian Kesenheimer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(29), 8805-8821 (2010-06-25)
A cobalt(I)-mediated convergent and asymmetric total synthesis of angucyclinones with an aromatic B ring has been developed. In the course of our research, we synthesized three naturally occurring anguclinone derivatives, namely, (+)-rubiginone B(2) (1), (-)-8-O-methyltetrangomycin (2), and (-)-tetrangomycin (3). By
Christian Kesenheimer et al.
Organic letters, 8(12), 2507-2510 (2006-06-02)
A stereoselective total synthesis of the natural antibiotic (-)-8-O-methyltetrangomycin 1 is reported. The essential steps for this convergent synthesis are the transformation of a geraniol epoxide into a chiral octadiyne derivative, which was converted into a triyne. The cobalt-mediated [2+2+2]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service