Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SHH0028

Sigma-Aldrich

Wrinkle Out Water Bath Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

form

liquid

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Application

Use in place of normal or distilled water for water bath when cutting paraffin sections and pick up wrinkle free ribbons and sections on the slide. Changes the water surface tension so tissue spreads evenly without wrinkles when using paraffin that is of lower quality than the polymer fortified paraffins on the market.

pictograms

FlameHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - STOT SE 2

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

54.9 °F

flash_point_c

12.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service