Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SEP120

Millipore

Seppro® Rat

LC10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

column D × L

12.7 mm × 79.0 mm

capacity

12 mg total protein loading(266 μL of rat plasma, average rat protein concentration 45 mg/mL)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Immunoaffinity partitioning of highly-abundant proteins (HAP) has proven to be one of the most effective approaches for overcoming the wide dynamic range of plasma protein concentration, as well as enabling the detection of low-abundant proteins (LAP). Avian polyclonal IgY (Immunoglobulin Yolk) antibodies have unique and advantageous features that allow highly-specific partitioning of protein mixtures. Sigma offers a novel Seppro® IgY-R7 column system, which takes you deeper into the rat plasma/serum proteome.

Proteins Partitioned:

Rat serum albumin
IgG
Fibrinogen
Transferrin
IgM
Haptoglobin
alpha1-Antitrypsin

Application

Seppro® Rat, LC10 (10 mL bed volume columns) is designed for use as a plasma/serum protein removal tool base upon avian antibody (IgY)-antigen immune affinity removal of seven highly abundant proteins (HAP) from rat plasma protein mixtures via liquid chromatography. Seppro® Rat, spin column is used to support the analysis of low abundance rat serum/plasma proteins.

Legal Information

Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • S4199Seppro® Dilution bufferSDS

  • S4324Seppro® Stripping BufferSDS

  • S4449Seppro® Neutralization BufferSDS

  • CLS8163Corning® Costar® Spin-X® centrifuge tube filters, cellulose acetate membrane, pore size 0.45 μm, non-sterileSDS

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service