Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SEP100

Millipore

Seppro® Supermix Mouse

LC5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

column D × L

12.7 mm × 39.5 mm

capacity

loading(Flow Through volume from IgY 14 LC10 (SEP090))

storage temp.

2-8°C

General description

After the removal of highly abundant proteins (HAP) with the Seppro® mouse depletion column (SEP110 or SEP 090), the next level of protein abundance, the moderately abundant proteins (MAP) remains an obstacle in accessing the (low abundant proteins) LAP, where the majority of biologically interesting and clinically important biomarkers reside. Seppro Supermix Mouse can further partition complex plasma/serum samples. This product is generated by immunizing chickens with mouse plasma proteins from which the 7 HAP have been removed. The resulting antibodies are mainly against MAP and or immunoreactive proteins. Both the collected flow-through fraction and eluted fraction can be used for downstream analysis.

Application

Seppro® Supermix, LC5 (5 mL column) is used as avian polyclonal IgY-based immunoaffinity follow-on separation of mouse serum moderately abundant proteins (MAP) from mouse serum low abundant proteins (LAP) after specific mouse serum highly-abundant proteins (HAP) have been removed using Seppro® mouse depletion column SEP10.[1][2]

Legal Information

Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • S4199Seppro® Dilution bufferSDS

  • S4324Seppro® Stripping BufferSDS

  • S4449Seppro® Neutralization BufferSDS

  • CLS8163Corning® Costar® Spin-X® centrifuge tube filters, cellulose acetate membrane, pore size 0.45 μm, non-sterileSDS

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin König et al.
Clinical biochemistry, 46(7-8), 652-655 (2013-01-23)
Investigation of the practicability and performance of a magnetic micro-particle based method for protein depletion of serum samples, preceding the quantitative analysis of small molecules by LC-MS/MS. A commercially available kit including a protein denaturation reagent and functionalized magnetic particles
Majlinda Kullolli et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 939, 10-16 (2013-10-05)
Human plasma is a commonly used diagnostic fluid in clinical chemistry. In-depth plasma proteomic analysis is performed to search for disease biomarkers, however the large dynamic range of protein abundance in plasma presents a substantial analytical challenge. Removal of abundant

Questions

1–4 of 4 Questions  
  1. My protein sample contains SDS.  Is this compatible with Seppro® Supermix Mouse?

    1 answer
    1. No. We do not recommend using protein samples containing SDS with Product No. SEP100.  SDS is a detergent and would be expected to affect adversely the activity of the conjugated antibody in terms of depletion.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  4. How many moderately abundant proteins (MAPs) does Product SEP100, Seppro® Supermix Mouse, deplete?

    1 answer
    1. As of this time, the exact number of depleted MAPs by SEP100 is not precisely defined.  This question is currently under investigation.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service