All inquiries on the Seppro technology are now to be referred to Sigma-Aldrich. Customers should not contact Genway for technical assistance.
Recommended Products
bed volume
5 mL
column D × L
12.7 mm × 39.5 mm
capacity
loading(Flow Through volume from IgY 14 LC10 (SEP040))
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
To tackle this challenge, we further developed our IgY columns system by establishing a SuperMix column to separate MAP from LAP. The SuperMix System was developed by immunizing chickens with a flow-through fraction of IgY12 or IgY14 column and constructing the column with affinity-purified IgY antibodies against the flow-through proteins of IgY12 or IgY14.
Application
Other Notes
Legal Information
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
I originally purchased my Seppro SuperMix LC5 column from Genway. Can I still contact Genway for technical support?
1 answer-
Helpful?
-
-
My protein sample contains SDS. Is this compatible with Product SEP060, Seppro® Supermix LC5?
1 answer-
No. We do not recommend using protein samples containing SDS with Product No. SEP060. SDS is a detergent and would be expected to affect adversely the activity of the conjugated antibody in terms of depletion.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
How does the storage temperature relate to shipping conditions?
1 answer-
The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service