Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SE120011

Sigma-Aldrich

Ochratoxin A ELISA Kit for Alcoholic Beverages

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

technique(s)

ELISA: suitable
competitive inhibition ELISA: suitable

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 0-0.4 ng/mL
standard curve range: 0.02-0.4 ng/mL

shipped in

wet ice

storage temp.

2-8°C

General description

The Sigma-Aldrich® Ochratoxin A Alcoholic beverages ELISA (Enzyme-Linked Immunosorbent Assay) kit is a solid phase direct enzyme immunoassay.
An antibody with high affinity to Ochratoxin A is coated onto polystyrene microwells. Standard or sample is added to the appropriate well and if Ochratoxin A is present it will bind to the coated antibody. Subsequently, Ochratoxin A bound to horseradish peroxidase (HRP) is added and binds to the antibody not already occupied by Ochratoxin A present in the standard or sample. After this incubation period, the contents of the wells are decanted, washed and HRP substrate is added which develops a blue color in the presence of enzyme. The intensity of the color is directly proportional to the amount of bound conjugate and inversely proportional to the amount of Ochratoxin A in the standard or sample. Therefore, as the concentration of Ochratoxin A in the sample or standard increases, the intensity of the blue color will decrease. The reaction is stopped by the addition of an acid solution which causes the blue color to change to yellow.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1

Storage Class

5.1B - Oxidizing hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service