Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SE120008

Sigma-Aldrich

Total Aflatoxin ELISA Kit Low Matrix (Qual.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

technique(s)

ELISA: suitable
competitive inhibition ELISA: suitable

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 0-0.1 ng/mL
standard curve range: 0.04-0.1 ng/mL

shipped in

wet ice

storage temp.

2-8°C

General description

The Sigma-Aldrich® Low Matrix Total Aflatoxin ELISA (Enzyme-Linked Immunosorbent Assay) kit is a solid phase direct competitive enzyme immunoassay. An aflatoxin specific antibody optimized to cross react with all four subtypes of aflatoxin (see cross-reactivity information), is coated to a polystyrene microwell. Toxins are extracted from a ground sample with either 80% methanol or 80% acetonitrile and after dilution, added to the appropriate well. If aflatoxin is present it will bind to the coated antibody. Subsequently, aflatoxin bound to horse-radish peroxidase (HRP) is added and binds to the antibody not already occupied by aflatoxin present in the sample or standard. After this incubation period, the contents of the wells are decanted, washed and an HRP substrate is added which develops a blue color in the presence of enzyme. The intensity of the color is directly proportional to the amount of bound conjugate and inversely proportional to the amount of aflatoxin in the standard or sample. Therefore, as the concentration of aflatoxin in the sample or standard increases, the intensity of the blue color will decrease. A solution containing a red dye is added to stop the enzyme reaction and simultaneously causes a color change leaving the bluest wells deep blue/purple and the lightest wells a mauve/pink, thus facilitating a visual qualitative estimate of Aflatoxin in the sample by comparing to the color of the standards.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2 - Eye Dam. 1 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class

5.1B - Oxidizing hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service