Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0256

Sigma-Aldrich

Tyrosine Specific Protein Kinase Inhibitor Peptide A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C111H168N30O35
Molecular Weight:
2482.70
UNSPSC Code:
12352200
NACRES:
NA.32

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Val-Ala-Pro-Ser-Asp-Ser-Ile-Gln-Ala-Glu-Glu-Trp-Tyr-Phe-Gly-Lys-Ile-Thr-Arg-Arg-Glu

Application

Tyrosine Specific Protein Kinase Inhibitor Peptide A (VAPSDSIQAEEWYFGKITRRE) is used in as a tyrosine-dependent protein kinase inhibitor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service