Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0243

Sigma-Aldrich

SS14 Tumor Inhibitor Analog

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$607.00

$607.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
$607.00

About This Item

Empirical Formula (Hill Notation):
C54H69N11O10S2
Molecular Weight:
1096.32
UNSPSC Code:
12352200
NACRES:
NA.32

$607.00


Usually ships in 2 weeks.

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Nal-Cys-Tyr-Trp-Lys-Val-Cys-Thr-NH2

Application

Angiopeptin [3-(2-naphthyl)-D-Ala-Cys-Tyr-D-Trp-Lys-Val-Cys-Thr-NH2 (BIM-23014 C)] is an endocrinologically-potent octapeptide analogue of somatostatin (SS14) with potential use as a growth hormone inhibitor and an antiproliferation agent.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service