Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0211

Sigma-Aldrich

PHI rat

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$229.00

$229.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
500 μG
$229.00

About This Item

Empirical Formula (Hill Notation):
C136H216N36O41
Molecular Weight:
3011.39
UNSPSC Code:
12352202
NACRES:
NA.32

$229.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

His-Ala-Asp-Gly-Val-Phe-Thr-Ser-Asp-Tyr-Ser-Arg-Leu-Leu-Gly-Gln-Ile-Ser-Ala-Lys-Lys-Tyr-Leu-Glu-Ser-Leu-Ile-NH2

Application

Peptide HI (PHI) is a member of the vasoactive intestinal peptide (VIP) family of peptides that include VIP, secretin, GRF, PHI, PHM, GIP, glucagon and oxyntomodulin.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service