Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0202

Sigma-Aldrich

Obestatin 11-23 mouse

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$88.50
5 MG
$304.00

$88.50


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$88.50
5 MG
$304.00

About This Item

Empirical Formula (Hill Notation):
C61H98N22O18
Molecular Weight:
1427.57
UNSPSC Code:
12352200
NACRES:
NA.77

$88.50


Usually ships in 2 weeks.

Request a Bulk Order

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥72%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Leu-Ser-Gly-Ala-Gln-Tyr-Gln-Gln-His-Gly-Arg-Ala-Leu-NH2

Application

Obestatin is a 23 amino acid peptide with potential anoretic anti-ghrelin properties. Obestatin (11-23) and obestatin (1-10) are often used in comparative analysis with full length obestatin (1-23).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service