Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0201

Sigma-Aldrich

Neuromedin U-8

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$118.00
5 MG
$356.00

$118.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$118.00
5 MG
$356.00

About This Item

Empirical Formula (Hill Notation):
C54H78N16O10
Molecular Weight:
1111.30
UNSPSC Code:
12352200
NACRES:
NA.32

$118.00


Usually ships in 2 weeks.

Request a Bulk Order

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2

Application

Neuromedin U is a neuropeptide that exists in various isoforms with a constant C-terminal heptapeptide, Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2. Neuromedin U cell signaling is mediated by two receptors, peripheral NmUR1 and central nervous system NmUR2. Neuromedin U-8 (NMU-8) (YFLFRPRN-NH2) is identical to the C-terminus of human Neuromedin U-25. NMU-8 and its analogues are used to study NMU cell signaling activity and specificity.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service