Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0164

Sigma-Aldrich

[Gln8,9]-Helodermin

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$407.00
1 MG
$607.00

$407.00


Usually ships in 2 weeks.


Select a Size

Change View
500 μG
$407.00
1 MG
$607.00

About This Item

Empirical Formula (Hill Notation):
C176H285N47O49
Molecular Weight:
3843.43
UNSPSC Code:
12352209
NACRES:
NA.32

$407.00


Usually ships in 2 weeks.

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥78%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

His-Ser-Asp-Ala-Ile-Phe-Thr-Gln-Gln-Tyr-Ser-Lys-Leu-Leu-Ala-Lys-Leu-Ala-Leu-Gln-Lys-Tyr-Leu-Ala-Ser-Ile-Leu-Gly-Ser-Arg-Thr-Ser-Pro-Pro-Pro-NH2

Application

Helodermin is a VIP-secretin-like peptide that acts via vasoactive intestinal peptide (VIP) receptors.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Robberecht et al.
Peptides, 9(2), 339-345 (1988-03-01)
The ability of VIP, PHI, secretin, helodermin, and seven N-terminally D-amino monosubstituted VIP and PHI analogs to occupy (125I)iodo-VIP labeled receptors and to activate adenylate cyclase was tested on human lung membranes purified by the method of Schachter et al.
M Hoshino et al.
FEBS letters, 178(2), 233-239 (1984-12-10)
The complete amino acid sequence of helodermin isolated from the venom of Gila monster was elucidated. The peptide was shown to be a basic pentatriacontapeptide amide: His-Ser-Asp-Ala-Ile-Phe-Thr-Gln-Gln-Tyr-Ser-Lys-Leu-Leu-Ala-Lys-Leu-Ala- Leu-Gln-Lys- Tyr-Leu-Ala-Ser-Ile-Leu-Gly-Ser-Arg-Thr-Ser-Pro-Pro-Pro-NH2. A high degree of sequence similarities to secretin/VIP/PHI/(PHM)/GRF from mammal and
A Vandermeers et al.
European journal of biochemistry, 164(2), 321-327 (1987-04-15)
Having previously isolated helodermin, the major peptide like vasoactive-intestinal-peptide and peptide-histidine-isoleucinamide, from the venom of the lizard Heloderma suspectum, we decided on a systematic exploration of all (VIP-PHI)-like peptides present in the venom of another lizard of the Helodermatidae family:
Mitsuteru Akahoshi et al.
The Journal of clinical investigation, 121(10), 4180-4191 (2011-09-20)
Mast cell degranulation is important in the pathogenesis of anaphylaxis and allergic disorders. Many animal venoms contain components that can induce mast cell degranulation, and this has been thought to contribute to the pathology and mortality caused by envenomation. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service