All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H34N6O5
Molecular Weight:
510.59
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥60%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Phe-Pro-Lys-pNA
Application
D-Phe-Pro-Lys-pNA may be used as a thrombin substrate and active site probe.
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service