Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0039

Sigma-Aldrich

Amyloid β 16-10 reverse human

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$150.00

$150.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$150.00

About This Item

Empirical Formula (Hill Notation):
C42H61N13O12
Molecular Weight:
940.01
UNSPSC Code:
12352200
NACRES:
NA.77

$150.00


Usually ships in 2 weeks.

Request a Bulk Order

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥72%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Lys-Gln-His-His-Val-Glu-Tyr

General description

Serves as a control for Aβ (10-16) as well as for antibody based techniques utilizing the Aβ (10-16) epitope (see product SCP0031).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service