Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0012

Sigma-Aldrich

Calmodulin Dependent Protein Kinase II 281-309

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$286.00

$286.00


Usually ships in 2 weeks.


Select a Size

Change View
1 MG
$286.00

About This Item

Empirical Formula (Hill Notation):
C146H254N46O39S3
Molecular Weight:
3374.06
UNSPSC Code:
12352202
NACRES:
NA.32

$286.00


Usually ships in 2 weeks.

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Met-His-Arg-Gln-Glu-Thr-Val-Asp-Cys-Leu-Lys-Lys-Phe-Asn-Ala-Arg-Arg-Lys-Leu-Lys-Gly-Ala-Ile-Leu-Thr-Thr-Met-Leu-Ala

Application

Calmodulin Dependent Protein Kinase II 281-309 [CaMK-(281-309)] is a CaMK-peptide used to probe interactions of the various regulatory domains of protein kinase II.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Colbran et al.
The Journal of biological chemistry, 263(34), 18145-18151 (1988-12-05)
Two peptide analogs of Ca2+/calmodulin-dependent protein kinase II (CaMK-(peptides)) were synthesized and used to probe interactions of the various regulatory domains of the kinase. CaMK-(281-289) contained only Thr286, the major Ca2+-dependent autophosphorylation site of the kinase (Schworer, C. M., Colbran
A Ishida et al.
The Journal of biological chemistry, 270(5), 2163-2170 (1995-02-03)
The active 30-kDa chymotryptic fragment of calmodulin-dependent protein kinase II (CaM kinase II), devoid of the autoinhibitory domain, and the enzyme, autothiophosphorylated at Thr286/Thr287, were much more labile than was the original native enzyme. They were markedly stabilized by synthetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service