Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0008

Sigma-Aldrich

[DArg0,Hyp3,Thi5,8,DPhe7]-Adamantaneacetyl Bradykinin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$88.50
5 MG
$304.00

$88.50


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$88.50
5 MG
$304.00

About This Item

Empirical Formula (Hill Notation):
C68H99N19O14S2
Molecular Weight:
1470.76
UNSPSC Code:
12352209
NACRES:
NA.32

$88.50


Usually ships in 2 weeks.

Request a Bulk Order

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ada-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-Phe-Thi-Arg

Application

Bradykinin, a nonapeptide, is a potent endothelium-dependent vasodilator that operates via bradykinin receptors B1 and B2. Adamantaneacetyl Bradykinin (NA-BK) and [DArg0,Hyp3,Thi5,8,DPhe7]-Adamantaneacetyl Bradykinin are used as a B2 bradykinin receptor antagonists.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galia K Soukhova-O'Hare et al.
Life sciences, 78(17), 1990-1997 (2005-11-18)
Endogenous bradykinin (BK) is an established mediator of pulmonary inflammation, yet its role in lung disease is unclear. In the rabbit, injecting BK into the lung parenchyma elicits reflex hyperpnea, tachypnea, hypotension, and bradycardia by stimulating pulmonary sympathetic afferents. To
W D Ehringer et al.
Inflammation, 21(3), 279-298 (1997-06-01)
alpha-Thrombin (AT) and bradykinin (BK) are endogenous mediators that are released during an inflammatory response, and could have a synergistic effect on endothelial permeability. Human umbilical vein endothelial cells (HUVEC) were grown on Transwell membranes and then tested for alterations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service