SCM122
AdipoMAX Differentiation Medium
Synonym(s):
Human MSC Adipogenesis Media, MSC adipocyte media, adipocyte differentiation media, adipogenesis media
About This Item
Recommended Products
shipped in
dry ice
storage temp.
−20°C
General description
AdipoMAX Differentiation Media is a ready-to-use mesenchymal stem cell adipogenesis differentiation media. AdipoMAX produces cells with elevated levels of lipids, analyzed by Oil-Red-O staining, after 3 weeks of differentiation compared to other adipogenesis differentiation media. AdipoMAX is an excellent research tool to study adipocyte differentiation, adipogenesis and adipocyte related metabolic diseases.
Application
Other Notes
Before use, thaw both materials and combine the dilution factor into the basal medium. The complete medium can be stored at 2-8°C for up to 3 weeks.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Frequently asked questions about mesenchymal stem cells including MSC derivation, expansion, differentiation and allogenic stem cell therapy.
Protocols
Information about mesenchyme, specifically mesenchymal stem cell procotols. Step-by-step cell culture protocols for mesenchymal stem cell (MSC) isolation, expansion and differentiation.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service