SCJ4600034
CF™ 620R succinimidyl ester
suitable for fluorescence, ≥90% (TLC)
Select a Size
$330.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$330.00
Notify Me
Get notified when this item is ready to ship via email.
Recommended Products
assay
≥90% (TLC)
form
solid
mol wt
~738 g/mol (free acid)
solubility
DMF: soluble
DMSO: soluble
H2O: soluble
ε (extinction coefficient)
115000 at 617 nm
absorbance ratio
A280/617 nm 0.45
fluorescence
λex 633 or 635 nm (Flow cytometry laser line)
λex 633 or 635 nm (Microscopy laser line)
λex 617 nm; λem 639 nm
suitability
suitable for fluorescence
shipped in
wet ice
storage temp.
−20°C
General description
Features and Benefits
Preparation Note
Reconstitution
Legal Information
Disclaimer
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service