Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SBPV09

Sigma-Aldrich

BCRP Human Vesicular HEK293 Transport Kit membrane based kit for Human BCRP Vesicular Transport Assays

Synonym(s):

SB-BCRP-HEK293-Predivez kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

recombinant

expressed in HEK 293 cells

form

liquid

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ABCG2(9429)

General description

SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVO′s PREDIVEZ Vesicular Transport Assay Kits are suitable for efflux transporter inhibition studies. The kits are designed to test the inhibitory effect of one or more test compounds on the vesicular transport of a probe substrate. In most cases a fluorescent probe substrate is included. Kits contain all reagents and solutions necessary to conduct the drug-transporter interaction assay of need.

Application

The PREDIVEZ Vesicular Transport Kit is a simple and powerful tool to investigate drug transporter interactions. It provides information on any interaction between the ABC transporter and the drug candidate that would affect the transport of the fluorescent or radiolabeled or cold.

PREDIVEZ vesicular transport kits are available in three sizes 3, 6 or 9 compound sizes. The kits contain the sufficient amount of reagents and membrane preparations needed for the testing of 3 compounds per 96-well plate. Each kit is accompanied by a CD that includes the assay protocol, an Excel spreadsheet with a data processing template, SDS and the inhibition curves with reference compounds.

Legal Information

PREDIVEZ is a trademark of Solvo Biotechnology

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

188.6 °F

flash_point_c

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service