Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAE0083

Sigma-Aldrich

Auresine

recombinant, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
EC Index Number:
3.4.24.75

recombinant

expressed in Pichia pastoris

form

lyophilized powder

specific activity

>500 units/mg protein

mol wt

14.4 kDa

storage condition

room temperature (at +4 °C (in Storage Buffer))

storage temp.

2-8°C

General description

Auresine is a portion of the zinc metalloprotease Staphylococcus aureus. The enzyme has glycyl-glycine endopeptidase activity and it specifically cleaves polyglycine crosslinks in the cellular wall of Staphylococcus species, including various strains of methicillin-resistant Staphylococcus aureus (MRSA). Auresine is thus useful for lysis of MRSA.

Application

Auresine can be used for isolation of protoplasts, nucleic acids, proteins, lipids and other components of Gram-positive bacterial cells. Auresine is particularly useful to lyse MRSA.. Thanks to its unique activity in non-physiological conditions (low conductivity and low temperatures), isolated components after auresine treatment, like nucleic acids and proteins, are not prone to degradation by released cellular enzymes.

Unit Definition

One unit will reduce the turbidity (A600) of suspension of S. aureus cells from 0.250 to 0.125 in 10 minutes in 50 mM glycine, pH 8.0 at 25°C in 6.0 mL reaction mixture.

Preparation Note

Auresine in soluble in water (20 mg/ml), yielding a clear solution. The product is active in 100 nM concentration (1.4 ug/mL) in low conductivity buffers (= 2 mS/cm) at temp. 0-40°C.

Storage and Stability

Auresine lyophilized powder should be stored at room temperature. Once dissolved in storage buffer, Auresine is stable at room temperature up to 4 months, at 4°C, -20°C, -80°C at least 2 years. Storage buffer: 20 mM Tris-HCl, pH 7.0, 200 mM NaCl, 10% glycerol.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service