Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

SAB5700473

Sigma-Aldrich

Anti-Phospho-VIM-S39 antibody produced in rabbit

Synonym(s):

CTRCT30, HEL113, VIM, Vimentin, vimentin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$314.00

$314.00


Check Cart for Availability


Select a Size

Change View
100 μL
$314.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$314.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53

species reactivity

human, mouse, rat

concentration

1.51 mg/ml

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

phosphorylation (pSer39)

Gene Information

human ... VIM(7431)

General description

This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataract.

Immunogen

A synthetic phosphorylated peptide around S39 of human VIM (NP_003371.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service