Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5700242

Sigma-Aldrich

Anti-Acetyl-Histone H3-K14 antibody produced in rabbit

Synonym(s):

H3.4, H3/g, H3FT, H3t, HIST1H3A, HIST3H3, Histone H3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$314.00

$314.00


Check Cart for Availability


Select a Size

Change View
100 μL
$314.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$314.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

15

species reactivity

mouse, rat, human

concentration

0.28 mg/ml

technique(s)

ChIP: 1:50-1:100
immunofluorescence: 1:500-1:1000
immunohistochemistry: 1:200-1:500
immunoprecipitation (IP): 1:200-1:500
western blot: 1:1000-1:3000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys14)

Gene Information

human ... H3-4(8290)

General description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3.

Immunogen

A synthetic acetylated peptide corresponding to residues surrounding K14 of human H3

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service