Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

SAB5700117

Sigma-Aldrich

Anti-Acetyl-Histone H2B-K15 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$458.00

$458.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$458.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$458.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

concentration

0.28 mg/ml

technique(s)

immunofluorescence: 1:50-1:100
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:200
western blot: 1:500-1:2000

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys15)

Gene Information

Immunogen

A synthetic acetylated peptide corresponding to residues surrounding K15 of human Histone H2B

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service