Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5700100

Sigma-Aldrich

Anti-DPYSL2 antibody produced in rabbit

Synonym(s):

CRMP-2, CRMP2, DHPRP2, DPYSL2, DRP-2, DRP2, N2A3, ULIP-2, ULIP2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$314.00

$314.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$314.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$314.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58
62

species reactivity

human

concentration

3.58 mg/ml

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPYSL2(1808)

General description

This gene encodes a member of the collapsin response mediator protein family. Collapsin response mediator proteins form homo- and hetero-tetramers and facilitate neuron guidance, growth and polarity. The encoded protein promotes microtubule assembly and is required for Sema3A-mediated growth cone collapse, and also plays a role in synaptic signaling through interactions with calcium channels. This gene has been implicated in multiple neurological disorders, and hyperphosphorylation of the encoded protein may play a key role in the development of Alzheimer′s disease. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 400-500 of human DPYSL2 (NP_001377.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service