Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5600140

Sigma-Aldrich

Anti-p63 (Tumor protein 63) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM329, purified immunoglobulin

Synonym(s):

Anti-AIS, Anti-B(p51A), Anti-B(p51B), Anti-EEC3, Anti-KET, Anti-LMS, Anti-NBP, Anti-OFC8, Anti-RHS, Anti-SHFM4, Anti-TP53CP, Anti-TP53L, Anti-TP73L, Anti-p40, Anti-p51, Anti-p53CP, Anti-p63, Anti-p73H, Anti-p73L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM329, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TP63(8626)

Specificity

This antibody reacts to human p63 (Tumor protein 63).

Immunogen

A peptide corresponding to the area surrounding the C-terminus of human p63 (Tumor protein 63).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Wang et al.
Nature communications, 12(1), 7006-7006 (2021-12-03)
Nicotine addiction and the occurrence of lymph node spread are two major significant factors associated with esophageal cancer's poor prognosis; however, nicotine's role in inducing lymphatic metastasis of esophageal cancer remains unclear. Here we show that OTU domain-containing protein 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service