SAB5600037
Anti-Phospho-Histone H2B (Ser14) antibody, Rabbit monoclonal
recombinant, expressed in HEK 293 cells, clone RM238, purified immunoglobulin
Synonym(s):
H2BpSer14, Histone H2B (phospho ser14)
Select a Size
$408.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$408.00
Notify Me
Get notified when this item is ready to ship via email.
recombinant
expressed in HEK 293 cells
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
RM238, monoclonal
recombinant monoclonal
form
buffered aqueous glycerol solution
species reactivity
human
concentration
~1 mg/mL
technique(s)
ELISA: 0.2- 1 μg/mL
immunoblotting: 0.5-2 μg/mL
immunocytochemistry: 1- 2 μg/mL
isotype
IgG
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
phosphorylation (pSer14)
Gene Information
human ... H2BC1(255626)
Specificity
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service